missing translation for 'onlineSavingsMsg'
Learn More

Silica, standard solution, Specpure™ SiO{2} 1000μg/mL

$150.95 - $150.95

Chemical Identifiers

MDL Number MFCD00011232
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42225AE
View Documents
Thermo Scientific Chemicals
042225AE
100 mL
Each for $150.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00011232

Specifications

Liquid
100 mL
Silica standard solution, SiO2 1000μg/mL
MFCD00011232
Specpure
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only