missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica Standard, Certified, 1000ppm ±10ppm (1mL = 1mg SiO2), LabChem™
Supplier: LabChem LC223901
Specifications
| Silica Standard | |
| Poly Bottle | |
| 99.53,0.47 | |
| 1g/mL | |
| H18Na2O12Si | |
| Na2SiO3·9H2O | |
| Soluble in water | |
| O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+] | |
| 284.197 | |
| 61639 | |
| Certified |
| 1000ppm (1mL = 1mg SiO2) | |
| 13517-24-3,7732-18-5 | |
| Passes Test | |
| Liquid | |
| 1000 ppm ±10 ppm (1mL = 1mg SiO2) | |
| Colorless | |
| PHIQPXBZDGYJOG-UHFFFAOYSA-N | |
| disodium;dioxido(oxo)silane;nonahydrate | |
| 500 mL | |
| 284.2 |
Chemical Identifiers
| 13517-24-3 | |
| 284.197 | |
| 61639 | |
| O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+] |
| H18Na2O12Si | |
| PHIQPXBZDGYJOG-UHFFFAOYSA-N | |
| disodium;dioxido(oxo)silane;nonahydrate |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature