missing translation for 'onlineSavingsMsg'
Learn More

Silica Standard, 1000 ppm SiO2 (467 ppm Si), Ricca Chemical

Supplier:  Ricca Chemical Company 67505

Catalog No. 67505

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Silica Standard
6834-92-0 , 7732-18-5
100.0, 0.53
Na2O3Si
[O-][Si](=O)[O-].[Na+].[Na+]
122.062
CHEBI:60720
Cubitainer
97.0, 0.51
1 mL = 1 mg SiO
NTHWMYGWWRZVTN-UHFFFAOYSA-N
disodium;dioxido(oxo)silane
23266
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents