missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel PS Drying Agent, Honeywell Fluka™

With indicator

$327.65 - $327.65

Chemical Identifiers

CAS 112926-00-8
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00011232
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60047561
View Documents
Honeywell-Fluka
78307250G
250 g Poly Bottle
Each for $327.65
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

112926-00-8
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
CHEBI:30563
O=[Si]=O
O2Si
MFCD00011232
24261
dioxosilane

Specifications

112926-00-8
250 g
MFCD00011232
Poly Bottle
O=[Si]=O
60.083
CHEBI:30563
Silica gel PS drying agent
Silica Gel
O2Si
NONH for all modes of transport
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.08g/mol
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'