missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel PS Drying Agent, Solstice

Catalog No. 60047561
Change view
Click to view available options
Quantity:
250 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047561 250 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60047561 Supplier Solstice Supplier No. 78307250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

With indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 112926-00-8
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00011232
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Material Silica Gel
Quantity 250 g
UN Number NONH for all modes of transport
Packaging Poly Bottle
Formula Weight 60.08g/mol
Chemical Name or Material Silica gel PS drying agent