missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, Indicating Beads Expose to atmosphere only when put into use., Reagents

Catalog No. p-200021013
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337076 10 kg
01337075 3 kg
01337074 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337076 Supplier Labchem Inc Supplier No. C230055010KG
Only null left

Silica Gel, Indicating Beads Expose to atmosphere only when put into use., Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1343-98-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Physical Form Solid
Color Blue
Material Silica Gel
Quantity 10 kg
Packaging Poly Pail
Specific Gravity 2.1
Chemical Name or Material Silica Gel, Indicating Beads Expose to atmosphere only when put into use.