missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, High-Purity Grade, MilliporeSigma™ Supelco™

Catalog No. 111012554
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012554 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012554 Supplier Merck Emd Millipore Supplier No. 607651KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation resulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is classified as below:a. Aquagel - pores are filled with waterb. Xerogel - by the process of evaporation, aqueous phase in the pores are removedc. Aerogel - solvent is removed by supercritical extraction

Specifications

CAS 112926-00-8
Physical Form Powder
MDL Number MFCD00011232
Melting Point >1600°C
Boiling Point 2230°C
Type Organic Standard
Quantity 1 kg
Grade High Purity