missing translation for 'onlineSavingsMsg'
Learn More

Silica 32 to 63 μm, 60A, EcoChrom™, Not Active, MP Biomedicals™

For Chromatography; Chemical Synthesis

$977.66 - $7204.85

Specifications

Content And Storage Store at Room Temperature.
Type Silica
Physical Form Powder
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Density 2g/cm3 at 20°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN02827
View Documents
MP Biomedicals Inc
0202827
2.5 kg
Each for $977.66
Only null left
 
ICN02826
View Documents
MP Biomedicals Inc
0202826
25 kg
Each for $7,204.85
Only null left
 
Description

Description

Specifications

Specifications

Store at Room Temperature.
Powder
2g/cm3 at 20°C
Silica
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
7631-86-9
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'