missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica 32 to 63 μm, 60A, EcoChrom™, Not Active, MP Biomedicals™
For Chromatography; Chemical Synthesis
Supplier: MP Biomedicals Inc 0202827
Specifications
| 2.5 kg | |
| Silica | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| 7631-86-9 |
| Store at Room Temperature. | |
| Powder | |
| 2g/cm3 at 20°C |