missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ Organic Reversed-Phase Kit

For your convenience, we have combined our most popular bulk reagents into the SiliaBond™ Organic Reversed-Phase Kit, so you can be prepared with material in hand for any application.

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase C8, C18, C18 nec, C18 WPD, Cyano, Phenyl
Technique Column Chromatography
Packaging Bulk
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01182172
View Documents
SiliCycle™
K32830B5G
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Our Kits are available in 5 g, 10 g, 25 g, 50 g or 100 g format.
  • Includes: C8, C18, C18 nec, C18 WPD, Cyano and Phenyl
  • All in 40 to 63 μm 60 Å
Specifications

Specifications

40 to 63 μm
C8, C18, C18 nec, C18 WPD, Cyano, Phenyl
Bulk
Keep dry.
Bulk Functionalized Irregular Silica Gel
Glass Gravity Columns
Irregular
60 Å
Column Chromatography
Reversed-Phase Kit (6 different phases)
SiliaBond
Silica Gel
Screening of reversed-phases
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'