missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ Organic Reversed-Phase Kit

Catalog No. 01182172
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01182172 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01182172 Supplier SiliCycle™ Supplier No. K32830B5G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For your convenience, we have combined our most popular bulk reagents into the SiliaBond™ Organic Reversed-Phase Kit, so you can be prepared with material in hand for any application.

  • Our Kits are available in 5 g, 10 g, 25 g, 50 g or 100 g format.
  • Includes: C8, C18, C18 nec, C18 WPD, Cyano and Phenyl
  • All in 40 to 63 μm 60 Å

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase C8, C18, C18 nec, C18 WPD, Cyano, Phenyl
Technique Column Chromatography
Packaging Bulk
Quantity 5 g
Sorbent Reversed-Phase Kit (6 different phases)
Content And Storage Keep dry.
Product Line SiliaBond
Type Bulk Functionalized Irregular Silica Gel
Material Silica Gel
For Use With (Equipment) Glass Gravity Columns
For Use With (Application) Screening of reversed-phases
Particle Shape Irregular
Show More Show Less