missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel Tosic Acid (SCX), 40 to 63 μm, 60 Å

Tosic Acid multifunctionlized gel is a chromatographic phase acting as a strong cation exchanger and an organic scavenger for trapping amines and anilines.

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase Tosic Acid (SCX)
Technique Column Chromatography
Grade Irregular Silica Gel
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01182317
View Documents
SiliCycle™
R60530B5G
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

SiliaBond™ Tosic Acid acts as a Chromatographic Phase and an Organic Scavenger for Bases. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. This gel has a great variety of formats for all purifications scales, from laboratory to process to plant scales. It is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment.

  • Strong cation exchangers of pKa <1, permanently negatively charged or pH independent; for catch and release purification of weak cations such as basic drugs, analgesics, biomolecules and water-soluble vitamins
  • Ionic Bonding and Catch and Release for Amines and Anilines
  • Great compatibility with a myriad of experimental conditions, solvents, pH and temperatures
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with a sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed
Specifications

Specifications

40 to 63 μm
Tosic Acid (SCX)
Irregular Silica Gel
500 m2/g
SiliaBond
Silica Gel
Bulk Scavenger in Post-reaction Mixture or Purification of Weak Basic Molecules
0.698 g/mL
HDPE Bottle
60 Å
Column Chromatography
Tosic Acid (Ethyltosic Acid, SCX)
Keep Dry
Bulk Functionalized Irregular Silica Gel
Bulk or Glass Gravity Columns
Irregular
Yes
Irregular
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'