Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel TMA Acetate (SAX-2) nec, 40 to 63 μm, 60 Å
Our TMA Acetate gel is a multifunctional chromatographic phase that acts as a strong anion exchanger with easily exchangeable acetate counter-ion.
Specifications
| Particle Size | 40 to 63 μm |
|---|---|
| Pore Size | 60 Å |
| Stationary Phase | TMA Acetate (SAX-2) nec |
| Technique | Column Chromatography |
| Grade | Irregular Silica Gel |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01182341
|
SiliCycle™
R66430B5G |
5 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
SiliaBond™ TMA Acetate (SAX-2) acts as a Chromatographic Phase and an Organic Scavenger for Acids. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. The gel has a great variety of formats for all purifications scales, from laboratory to process to plant scales. It is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and offers great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
- Strong anion exchangers with easily exchangeable acetate counter-ion - more than chloride ion - for compounds with pKa <5 such as carboxylic acids
- Ionic Bonding and Catch and Release for Acids and Acidic Phenols
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with a sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
| 40 to 63 μm | |
| TMA Acetate (SAX-2) nec | |
| Irregular Silica Gel | |
| 500 m2/g | |
| SiliaBond | |
| Silica Gel | |
| Bulk scavenger in post-reaction mixture or purification of weak acidic molecules | |
| 0.665 g/mL | |
| HDPE Bottle |
| 60 Å | |
| Column Chromatography | |
| TMA Acetate (Tributylammonium Chloride, SAX-2) nec | |
| Keep Dry | |
| Bulk Functionalized Irregular Silica Gel | |
| Bulk or Glass Gravity Columns | |
| Irregular | |
| No | |
| Irregular |