missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel TMA Acetate (SAX-2) nec, 40 to 63 μm, 60 Å
Description
SiliaBond™ TMA Acetate (SAX-2) acts as a Chromatographic Phase and an Organic Scavenger for Acids. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. The gel has a great variety of formats for all purifications scales, from laboratory to process to plant scales. It is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and offers great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
- Strong anion exchangers with easily exchangeable acetate counter-ion - more than chloride ion - for compounds with pKa <5 such as carboxylic acids
- Ionic Bonding and Catch and Release for Acids and Acidic Phenols
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with a sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
Specifications
| Particle Size | 40 to 63 μm |
| Pore Size | 60 Å |
| Stationary Phase | TMA Acetate (SAX-2) nec |
| Technique | Column Chromatography |
| Grade | Irregular Silica Gel |
| Quantity | 5 g |
| Sorbent | TMA Acetate (Tributylammonium Chloride, SAX-2) nec |
| Surface Area (Metric) | 500 m2/g |
| Content And Storage | Keep Dry |
| Product Line | SiliaBond |
| Show More |