missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel Propylsulfonic Acid (SCX-2) nec, 40 to 63 μm, 60 Å

Propylsulfonic Acid (SCX-2) nec is a multifunctional gel which acts as a strong cation exchanger for catch and release of weak cations and removes Amines and Anilines.

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase Propylsulfonic Acid (SCX-2) nec
Technique Column Chromatography
Grade Irregular Silica Gel
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01182285
View Documents
SiliCycle™
R51430B5G
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

SiliaBond™ Propylsulfonic Acid (SCX-2) acts as a Chromatographic Phase and an Organic Scavenger for Bases. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. This gel provides a great variety of formats for all purifications scales, from laboratory to process to plant scales. It is successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and has great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • Strong cation exchangers of pKa <1, permanently negatively charged and pH independent; for catch and release purification of weak cations such as basic drugs, analgesics, biomolecules and water-soluble vitamins
  • Ionic Bonding and Catch and Release for Amines and Anilines
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed
Specifications

Specifications

40 to 63 μm
Propylsulfonic Acid (SCX-2) nec
Irregular Silica Gel
500 m2/g
SiliaBond
Silica Gel
Post-reaction Mixture or Extraction of Weak Basic Molecules, Catch and Release of Amines
0.728 g/mL
HDPE Bottle
60 Å
Column Chromatography
Propylsulfonic Acid (SCX-2) nec
Keep Dry
Bulk Functionalized Irregular Silica Gel
Bulk or Glass Gravity Columns
Irregular
No
Irregular
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'