missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel Propylsulfonic Acid (SCX-2) nec, 40 to 63 μm, 60 Å

Catalog No. 01182285
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01182285 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01182285 Supplier SiliCycle™ Supplier No. R51430B5G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Propylsulfonic Acid (SCX-2) nec is a multifunctional gel which acts as a strong cation exchanger for catch and release of weak cations and removes Amines and Anilines.

SiliaBond™ Propylsulfonic Acid (SCX-2) acts as a Chromatographic Phase and an Organic Scavenger for Bases. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. This gel provides a great variety of formats for all purifications scales, from laboratory to process to plant scales. It is successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and has great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • Strong cation exchangers of pKa <1, permanently negatively charged and pH independent; for catch and release purification of weak cations such as basic drugs, analgesics, biomolecules and water-soluble vitamins
  • Ionic Bonding and Catch and Release for Amines and Anilines
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase Propylsulfonic Acid (SCX-2) nec
Technique Column Chromatography
Grade Irregular Silica Gel
Quantity 5 g
Sorbent Propylsulfonic Acid (SCX-2) nec
Surface Area (Metric) 500 m2/g
Content And Storage Keep Dry
Product Line SiliaBond
Type Bulk Functionalized Irregular Silica Gel
Material Silica Gel
For Use With (Equipment) Bulk or Glass Gravity Columns
For Use With (Application) Post-reaction Mixture or Extraction of Weak Basic Molecules, Catch and Release of Amines
Physical Form Irregular
Density 0.728 g/mL
Endcapped No
Packing Material HDPE Bottle
Particle Shape Irregular
Show More Show Less