missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel Propylsulfonic Acid (SCX-2), 40 to 63 μm, 60 Å

Our Propylsulfonic Acid is a strong cation exchanger (pKa <1) and a powerful Organic Scavenger for the removal of amines and anilines.

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase Propylsulfonic Acid (SCX-2)
Technique Column Chromatography
Grade Irregular Silica Gel
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01182277
View Documents
SiliCycle™
R51230B5G
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

SiliaBond™ Propylsulfonic Acid (SCX-2) acts as a Chromatographic Phase and an Organic Scavenger for Bases. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Our propylsulfonic acid offers a great variety of formats for all purifications scales, from laboratory to process to plant scales, as well as great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • Strong cation exchangers of pKa <1, permanently negatively charged and pH independent
  • For catch and release purification of weak cations such as basic drugs, analgesics, biomolecules and water-soluble vitamins
  • Ionic Bonding and Catch and Release for Amines and Anilines
  • Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed
Specifications

Specifications

40 to 63 μm
Propylsulfonic Acid (SCX-2)
Irregular Silica Gel
500 m2/g
SiliaBond
Silica Gel
Bulk Scavenger in Post-reaction Mixture or Purification of Weak Basic Molecules
0.728 g/mL
HDPE Bottle
60 Å
Column Chromatography
Propylsulfonic Acid (SCX-2)
Keep Dry
Bulk Functionalized Irregular Silica Gel
Bulk or Glass Gravity Columns
Irregular
Yes
Irregular
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'