missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel Propylsulfonic Acid (SCX-2), 40 to 63 μm, 60 Å
Description
SiliaBond™ Propylsulfonic Acid (SCX-2) acts as a Chromatographic Phase and an Organic Scavenger for Bases. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Our propylsulfonic acid offers a great variety of formats for all purifications scales, from laboratory to process to plant scales, as well as great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
- Strong cation exchangers of pKa <1, permanently negatively charged and pH independent
- For catch and release purification of weak cations such as basic drugs, analgesics, biomolecules and water-soluble vitamins
- Ionic Bonding and Catch and Release for Amines and Anilines
- Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
Specifications
| Particle Size | 40 to 63 μm |
| Pore Size | 60 Å |
| Stationary Phase | Propylsulfonic Acid (SCX-2) |
| Technique | Column Chromatography |
| Grade | Irregular Silica Gel |
| Quantity | 5 g |
| Sorbent | Propylsulfonic Acid (SCX-2) |
| Surface Area (Metric) | 500 m2/g |
| Content And Storage | Keep Dry |
| Product Line | SiliaBond |
| Show More |