Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel Piperidine (PIP), 40 to 63 μm, 60 Å
Our Piperidine is designed to be easily be filtered out once the reaction is completed and is a powerful reagent for Knoevenagel condensations.
Supplier:  SiliCycle™ R71530B100G
 View more versions of this product
Catalog No. 01184407
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
SiliaBond Piperidine (Si-PIP) is a general purpose tertiary amine base to scavenge acids, thereby avoiding salt elimination problems.
  • Also used as base catalyst for Knoevenagel condensation
Specifications
Specifications
Irregular
HDPE Bottle
500 m2/g
100 g
Yes
Irregular
60 Å
Piperidine (PIP)
Post-reaction Mixture or in Flash Chromatography
40 to 63 μm
0.671 g/mL
Bulk Functionalized Irregular Silica Gel
Bulk or Glass Gravity Columns
Piperidine
SiliaBond
Product Suggestions
Product Suggestions