missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel Piperidine (PIP), 40 to 63 μm, 60 Å

Catalog No. p-7238156
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
50 g
100 g
250 g
500 g
1 kg
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01184404 5 g
01184433 10 g
01184405 25 g
01184406 50 g
01184407 100 g
01184408 250 g
01184434 500 g
01184422 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01184404 Supplier SiliCycle™ Supplier No. R71530B5G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Our Piperidine is designed to be easily be filtered out once the reaction is completed and is a powerful reagent for Knoevenagel condensations.

SiliaBond Piperidine (Si-PIP) is a general purpose tertiary amine base to scavenge acids, thereby avoiding salt elimination problems.

  • Also used as base catalyst for Knoevenagel condensation

Specifications

Physical Form Irregular
For Use With (Application) Post-reaction Mixture or in Flash Chromatography
Packaging HDPE Bottle
Particle Size 40 to 63 μm
Specific Surface Area 500 m2/g
Density 0.671 g/mL
Quantity 5 g
Product Type Bulk Functionalized Irregular Silica Gel
Endcapped Yes
For Use With (Equipment) Bulk or Glass Gravity Columns
Particle Shape Irregular
Phase Piperidine
Pore Size 60 Å
Product Line SiliaBond
Sorbent Piperidine (PIP)
Show More Show Less