missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel Piperidine (PIP), 40 to 63 μm, 60 Å
Our Piperidine is designed to be easily be filtered out once the reaction is completed and is a powerful reagent for Knoevenagel condensations.
Supplier: SiliCycle™ R71530B25G
Description
SiliaBond™ Piperidine (Si-PIP) is a general purpose tertiary amine base to scavenge acids, thereby avoiding salt elimination problems.
- Also used as base catalyst for Knoevenagel condensation
Specifications
| Irregular | |
| HDPE Bottle | |
| 500 m2/g | |
| 25 g | |
| Yes | |
| Irregular | |
| 60 Å | |
| Piperidine (PIP) |
| Post-reaction Mixture or in Flash Chromatography | |
| 40 to 63 μm | |
| 0.671 g/mL | |
| Bulk Functionalized Irregular Silica Gel | |
| Bulk or Glass Gravity Columns | |
| Piperidine | |
| SiliaBond |