Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel Carbonate (CO3), 40 to 63 μm, 60 Å
Our Carbonate Silica Gel is a powerful scavenger for the removal of acids, acidic phenols and boronic acids.
Specifications
| Chemical Name or Material | Bulk Functionalized Irregular Silica Gel |
|---|---|
| Physical Form | Irregular |
| Packaging | HDPE Bottle |
| Product Line | SiliaBond |
| Density | 0.608 g/mL |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01184000
|
SiliCycle™
R66030B100G |
100 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
SiliaBond™ Carbonate acts as an Organic Scavenger for Acids (Ionic Bonding and Catch and Release for Acids, Acidic Phenols and Boronic Acids). You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. The gel is great for a variety of formats for all purifications scales, from laboratory to process to plant scales. It is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and offers great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
| Bulk Functionalized Irregular Silica Gel | |
| HDPE Bottle | |
| 0.608 g/mL |
| Irregular | |
| SiliaBond |