missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ Functionalized Silica Gel Carbonate (CO3), 40 to 63 μm, 60 Å

Catalog No. 01184000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01184000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01184000 Supplier SiliCycle™ Supplier No. R66030B100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Our Carbonate Silica Gel is a powerful scavenger for the removal of acids, acidic phenols and boronic acids.

SiliaBond™ Carbonate acts as an Organic Scavenger for Acids (Ionic Bonding and Catch and Release for Acids, Acidic Phenols and Boronic Acids). You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. The gel is great for a variety of formats for all purifications scales, from laboratory to process to plant scales. It is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and offers great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed

Specifications

Chemical Name or Material Bulk Functionalized Irregular Silica Gel
Physical Form Irregular
Quantity 100 g
Packaging HDPE Bottle
Product Line SiliaBond
Density 0.608 g/mL