missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ C8 (11% C) Monomeric, 40 to 63 μm, 60 Å

SiliaBond™ C8 (11% C) Monomeric, 40 - 63 μm, 60 Å is a chromatographic phase for SPE or Flash/Gravity chromatography.

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase C8 Monomeric
Technique Column Chromatography
Sorbent C18 (Octadecyl, ODS)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01182205
View Documents
SiliCycle™
R30830B5G
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Less retention compared to C18.
  • For highly hydrophobic pesticides, small peptides and large molecule drugs.
Specifications

Specifications

40 to 63 μm
C8 Monomeric
C18 (Octadecyl, ODS)
SiliaBond
Silica Gel
Purification of large and highly hydrophobic molecules
60 Å
Column Chromatography
Keep dry.
Bulk Functionalized Irregular Silica Gel
Glass Gravity Columns
11%
Videos
SDS
Documents

Documents