missing translation for 'onlineSavingsMsg'
Learn More

SiliCycle™ SiliaBond™ C8 (11% C) Monomeric, 40 to 63 μm, 60 Å

Catalog No. 01182205
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01182205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01182205 Supplier SiliCycle™ Supplier No. R30830B5G

SiliaBond™ C8 (11% C) Monomeric, 40 - 63 μm, 60 Å is a chromatographic phase for SPE or Flash/Gravity chromatography.

  • Less retention compared to C18.
  • For highly hydrophobic pesticides, small peptides and large molecule drugs.

Specifications

Particle Size 40 to 63 μm
Pore Size 60 Å
Stationary Phase C8 Monomeric
Technique Column Chromatography
Quantity 5 g
Sorbent C18 (Octadecyl, ODS)
Content And Storage Keep dry.
Product Line SiliaBond
Type Bulk Functionalized Irregular Silica Gel
Material Silica Gel
For Use With (Equipment) Glass Gravity Columns
For Use With (Application) Purification of large and highly hydrophobic molecules
Carbon Load 11%
Show More Show Less