Learn More
Ambion™ Silencer™ Pre-Designed siRNA DFS Item
Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.
Supplier:  Ambion™ AM16708
 View more versions of this product
Catalog No. AM16708
Only null left
Explore available promotions
Description
Description
Silencer Predesigned siRNAs are high-quality, synthetic small interfering RNAs (siRNAs) designed for gene silencing applications in mammalian cells. This cost-effective RNA interference tool delivers reliable gene knockdown performance while maintaining economic accessibility for research applications. Our siRNA products are meticulously crafted to help ensure high efficiency and specificity, enabling researchers to achieve reliable and reproducible gene knockdown results. Silencer Predesigned siRNAs are unmodified synthetic RNA duplexes with a 19-mer duplex and two-nucleotide 3' overhangs.
Key features include:
  • High silencing efficiency—demonstrates good knockdown performance with optimized target recognition for effective gene expression suppression
  • Enhanced specificity—algorithm designed for precise gene targeting to minimize off-target effects and help ensure selective gene silencing
  • Guaranteed performance—two out of three predesigned siRNAs will result in >70% gene knockdown. Validated designs available for many common genes.
  • Cost-effective solution—exceptional value for research applications without compromising performance quality
Streamlined workflow for rapid results
Utilizing our gene knockdown technology is straightforward and efficient. Simply order siRNAs for the desired gene targets, deliver to the cells, and detect knockdown efficiency and validate the resulting phenotype. Take advantage of our validated designs for better confidence in your experiments. This streamlined process allows for rapid experimental progress and accelerated discovery in gene function studies.
Specifications
Specifications
Silencer™ Select siRNAs are the result of recent improvements in siRNA design combined with novel chemical modifications yielding unrivaled knockdown efficiency and specificity. Our best performing siRNA for cell culture work. Silencer™ siRNAs are unmodified 21 nucleotide siRNAs that have been designed with an older, yet rigorously tested design algorithm. A lower cost alternative to Silencer™ Select siRNAs, but without some of the performance enhancements.
96-well Plate, 384-well Plate
Silencer Pre-Designed siRNA
Silencer, Ambion
Standard
Silencer Pre-Designed siRNA, 5nmol, Incorporate the Latest Improvements in siRNA Design, Off-Target Effect Prediction Algorithms, and Chemistry, Guaranteed-to-Silence Based on their Proven Design
siRNA
Unlabeled
Silencer™ Pre-Designed siRNA
5 nmol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.