Learn More
Ambion™ Silencer™ Pre-Designed siRNA
Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.
$311.00 - $500.00
| Catalog Number | Mfr. No. | Product Type | Purity | Label or Dye | Quantity | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Purity | Label or Dye | Quantity | Price | Quantity & Availability | ||
AM16708
|
Ambion™
AM16708 |
Silencer™ Pre-Designed siRNA | Standard | Unlabeled | 5 nmol |
Each for $311.00
|
|
||
AM16706
|
Ambion™
AM16706 |
Silencer™ Pre-Designed Standard siRNA | Standard | Unlabeled | 40 nmol |
Each for $500.00
|
|
||
Description
Silencer Predesigned siRNAs are high-quality, synthetic small interfering RNAs (siRNAs) designed for gene silencing applications in mammalian cells. This cost-effective RNA interference tool delivers reliable gene knockdown performance while maintaining economic accessibility for research applications. Our siRNA products are meticulously crafted to help ensure high efficiency and specificity, enabling researchers to achieve reliable and reproducible gene knockdown results. Silencer Predesigned siRNAs are unmodified synthetic RNA duplexes with a 19-mer duplex and two-nucleotide 3' overhangs.
Key features include:
- High silencing efficiency—demonstrates good knockdown performance with optimized target recognition for effective gene expression suppression
- Enhanced specificity—algorithm designed for precise gene targeting to minimize off-target effects and help ensure selective gene silencing
- Guaranteed performance—two out of three predesigned siRNAs will result in >70% gene knockdown. Validated designs available for many common genes.
- Cost-effective solution—exceptional value for research applications without compromising performance quality
Streamlined workflow for rapid results
Utilizing our gene knockdown technology is straightforward and efficient. Simply order siRNAs for the desired gene targets, deliver to the cells, and detect knockdown efficiency and validate the resulting phenotype. Take advantage of our validated designs for better confidence in your experiments. This streamlined process allows for rapid experimental progress and accelerated discovery in gene function studies.
Specifications
| siRNA |
For Research Use Only. Not for use in diagnostic procedures.