missing translation for 'onlineSavingsMsg'
Learn More

Promega SignaTECT™ Protein Kinase Assay Systems

Catalog No. PRV8161
Encompass
Change view
Click to view available options
Target:
Calcium/Calmodulin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Target
PRV8161 Calcium/Calmodulin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV8161 Supplier Promega Supplier No. V8161
Only null left

Contain proprietary SAM2 Biotin Capture Membrane for higher binding capacity and specificity

  • Produces lower backgrounds and higher signal-to-noise ratios as compared to traditional P81 phosphocellulose methods
  • Perforated and numbered membrane allows measurement of up to 96 kinase reactions; ideal for kinetic studies
  • Assay requires little “hands-on” manipulation and is complete in under an hour
  • Can be used in a variety of buffer/reaction conditions including cell extracts
  • Each assay system contains highly specific biotinylated peptide substrates for the appropriate kinase as well as necessary reaction components
  • PTK assay system also contains two proprietary peptide substrates

User must supply [γ-32P] ATP

U.S. Patents 6,066,462 and 6,348,310 and other patents pending.

Specifications

Target Calcium/Calmodulin
For Use With (Application) Can be used in a variety of Buffer or reaction conditions including cell extracts
Includes ATP, BSA, SAM2 Biotin capture membrane, Termination Buffer, CaM KII Biotinylated Peptide Substrate, reaction control, activation Buffer
Content And Storage -20°C
Product Type Protein Kinase Assay
Quantity 96 Reactions