missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Pseudo™ Narcotic Scent (Cocaine Formulation Powder), MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | Sigma Pseudo |
| Physical Form | Powder |
| MDL Number | MFCD00165842 |
| Type | Standard |
| Quantity | 100 g |
Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC