missing translation for 'onlineSavingsMsg'
Learn More

Sigma Pseudo™ Corpse Scent Kit, MilliporeSigma™ Supelco™

Catalog No. 111004739
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004739 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004739 Supplier Merck Emd Millipore Supplier No. PSCI1KT

Specifications

Product Line Sigma Pseudo
Physical Form Neat
MDL Number MFCD00165840
Type Standard
Quantity 1 Kit

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC