missing translation for 'onlineSavingsMsg'
Learn More

Sigma-Aldrich™ 1D-1-O-Butyryl-4,6-O-dibenzoyl-myo-inositol 98, MilliporeSigma™ Supelco™

Catalog No. 111013226
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013226 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013226 Supplier Merck Emd Millipore Supplier No. 20787

Specifications

CAS 153265-90-8
Product Line Sigma-Aldrich
Synonym Hamilton(R) Syringe 702 RN 25 UL
Type Standard
Quantity 1 Ea.

Hamilton is a registered trademark of Hamilton Co.