missing translation for 'onlineSavingsMsg'
Learn More

Sialyl Lewis X-Lactose 97.0+%, TCI America™

Catalog No. p-7135962
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08491MG 1 mg
S08495MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08491MG Supplier TCI America Supplier No. S08491MG

Chemical Identifiers

CAS 127923-85-7
Molecular Formula C43H72N2O33
Molecular Weight (g/mol) 1145.03
PubChem CID 133554220

Specifications

Color White
Quantity 1 mg
Synonym sialyl lewis x-lactose, neu5ac alpha 2-3 gal beta 1-4 fuc alpha 1-3 glcnac beta 1-3 gal beta 1-4 glc
InChI Key QLSXTAKLCDGTLR-UHFFFAOYNA-N
Formula Weight 1145.03
Percent Purity >97.0% (HPLC)
Physical Form Crystalline Powder
Recommended Storage <0°C