accessibility menu, dialog, popup

UserName

CH3OOOOOOHOHOOOHHOOHHNOH3CHOOOHHOOHOOHOOOOHOHOHOHOHOHHNOH3COHHOHO

CAS RN 127923-85-7

IUPAC Name: 2-({2-[(6-{[3,5-dihydroxy-2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-4-yl]oxy}-5-acetamido-2-(hydroxymethyl)-4-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]oxan-3-yl)oxy]-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl}oxy)-5-acetamido-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Synonyms: 2-({2-[(6-{[3,5-dihydroxy-2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-4-yl]oxy}-5-acetamido-2-(hydroxymethyl)-4-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]oxan-3-yl)oxy]-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl}oxy)-5-acetamido-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Molecular Weight (g/mol): 1145.03
Molecular Formula: C43H72N2O33
InChi Key: QLSXTAKLCDGTLR-UHFFFAOYNA-N
SMILES: CC1OC(OC2C(NC(C)=O)C(OC3C(O)C(CO)OC(OC4C(CO)OC(O)C(O)C4O)C3O)OC(CO)C2OC2OC(CO)C(O)C(OC3(CC(O)C(NC(C)=O)C(O3)C(O)C(O)CO)C(O)=O)C2O)C(O)C(O)C1O
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)