missing translation for 'onlineSavingsMsg'
Learn More

Sialyl Lewis X-Lactose 97.0+%, TCI America™

Catalog No. S08495MG
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08495MG 5 mg
S08491MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08495MG Supplier TCI America Supplier No. S08495MG

Chemical Identifiers

CAS 127923-85-7
Molecular Formula C43H72N2O33
Molecular Weight (g/mol) 1145.03
PubChem CID 133554220
IUPAC Name (2S,4R,5R)-5-acetamido-2-[(2S,3S,5S)-2-[(3S,5S,6S)-5-acetamido-6-[(3S,5S,6S)-3,5-dihydroxy-2-(hydroxymethyl)-6-[(3S,5S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-4-yl]oxy-2-(hydroxymethyl)-4-[(2S,3S,5S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan
SMILES CC1C(C(C(C(O1)OC2C(C(OC(C2OC3C(C(C(C(O3)CO)O)OC4(CC(C(C(O4)C(C(CO)O)O)NC(=O)C)O)C(=O)O)O)CO)OC5C(C(OC(C5O)OC6C(OC(C(C6O)O)O)CO)CO)O)NC(=O)C)O)O)O

Specifications

Color White
Quantity 5 mg
Synonym Neu5Ac alpha(2-3)Gal beta(1-4)[Fuc alpha(1-3)]GlcNAc beta(1-3)Gal beta(1-4)Glc
InChI Key QLSXTAKLCDGTLR-QLUWDBCNSA-N
Formula Weight 1145.03
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sialyl Lewis X-Lactose
TSCA No
Recommended Storage Freezer