missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Shrimp Alkaline Phosphatase (SAP)

Catalog No. 78390500UN
Encompass
Change view
Click to view available options
Quantity:
1000 Units
500 Units
5000 Units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78390500UN 500 Units
783901000UN 1000 Units
783905000UN 5000 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 78390500UN Supplier Applied Biosystems™ Supplier No. 78390500UN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Shrimp Alkaline Phosphatase (SAP) is a high specific activity, heat-labile alkaline phosphatase purified from a recombinant source and originally isolated from Pandalus borealis (arctic shrimp).

  • 100% heat-inactivated in 5 min at 65°C
  • Significantly improved storage stability at lower temperatures
  • Very high specific activity
  • Removes 5'-phosphates from DNA, RNA, dNTPs, and proteins
  • Purified from a recombinant source
  • May be added directly to restriction enzyme digests
  • No vector purification necessary
  • Requires no supplemental zinc or other additives for activity
  • Works direct in many different buffers
  • Easy treatment of unincorporated dNTPs in PCR products prior to DNA sequencing or SNP analysis

Order Info

500 Units

Specifications

Concentration 1 unit/μL
Enzyme Alkaline Phosphatase
For Use With (Application) Dephosphorylation of DNA/RNA
Quantity 500 Units

For Research Use Only. Not for use in diagnostic procedures.