SHOWA™ Nitri-Pro™ Gloves
Ideal replacements for heavy cotton or leather work gloves
Manufacturer: SHOWA™ 716610
Description
For handling rough, abrasive materials such as castings, concrete blocks, sheet metal, extrusions and drywall. Extra-tough nitrile coating with Sanitized™ jersey liners for excellent resistance to cuts, punctures, snags and abrasions, as well as oil and grease.
- Fully coated, smooth finish style provides maximum protection and good dry grip and flexibility
- Palm coated, smooth finish style provides a good dry grip with even more flexibility
- All styles can be laundered and reused
Compliance
Styles comply with 21CFR170-199.
Specifications
Full | |
Smooth | |
heavy duty handling of castings, cores and fabricated Metals, brick, tile, plywood, drywall | |
Cotton Jersey | |
Navy |
Launder at 40°C using a neutral detergent | |
Reinforced safety | |
Nitrile | |
10 |