missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes: Toluene

Catalog No. 18556J
Change view
Click to view available options
Measurement Range:
100 to 1800
5–300ppm
50 to 400
Tube Type:
Toluene 100/a
Toluene 5/b
Toluene 50/a
Humidity Range:
5 to 12mg/L
Less than30mg/L
Not Applicable
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Tube Type Humidity Range
18556J 5–300ppm Toluene 5/b Not Applicable
18556L 100 to 1800 Toluene 100/a Less than30mg/L
18556K 50 to 400 Toluene 50/a 5 to 12mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18556J Supplier Dräger™ Supplier No. 8101661

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detector tubes

Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.

Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.

Specifications

Components Measured Toluene
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range Not Applicable
Measurement Range 5–300ppm
No. of Strokes 2/10
Quantity 10
Response Time 1 to 5 min.
Temperature Range (English) 41°F to 77°F
Temperature Range (Metric) 2° to 40°C
Tube Type Toluene 5/b
Show More Show Less