missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes: Chlorine

Catalog No. 17985207
Change view
Click to view available options
Measurement Range:
0.2 to 30
50 to 500
Tube Type:
Chlorine 0.2/a
Chlorine 50/a
Humidity Range:
Less than40mg/L
Not Applicable
No. of Strokes:
1
10/1
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Tube Type Humidity Range No. of Strokes
17985207 50 to 500 Chlorine 50/a Less than40mg/L 1
17985206 0.2 to 30 Chlorine 0.2/a Not Applicable 10/1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17985207 Supplier Dräger™ Supplier No. CH20701

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detector tubes

Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.

Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.

Specifications

Components Measured Chlorine
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range Less than40mg/L
Measurement Range 50 to 500
No. of Strokes 1
Quantity 10
Response Time 20 sec.
Temperature Range (English) 50°F to 104°F
Temperature Range (Metric) 10° to 40°C
Tube Type Chlorine 50/a
Show More Show Less