Learn More
Poltex Short Serological Pipette Holder
To organize pipettes and is perfect for under counter, ergonomic, or in hood (biosafety , PCR , tissue culture) placement.
Supplier:  Poltex SRPIPSHRTRPOK
Encompass
 View more versions of this product
Catalog No. 22000227
Only null left
Description
Description
  • Made from durable break and alcohol resistant PETG
  • Non-slip rubber feet provide stable base
  • Removable shelves for easy cleaning
  • Available with or without pockets
Specifications
Chemical Identifiers
Not Autoclavable
Serological Pipets (4 Types)
Non-sterile
10 in.
25.4 cm
Clear
PETG
Serological Pipettes
10 in.
25.4 cm
Specifications
Not Autoclavable
Serological Pipets (4 Types)
Non-sterile
10 in.
8.25 in.
Serological Pipet Holder with Right Pockets
25.4 cm
Clear
PETG
Serological Pipettes
10 in.
20.9 cm
25.4 cm
Product Suggestions
Product Suggestions