Learn More
Synthware™ Short Path Distillation Apparatus- Microscale
Specifications
Specifications
| For Use With (Application) | Organic Synthesis |
| Type | Distillation Apparatus |
| Material | Borosilicate Glass |
| For Use With (Equipment) | 25 mm Immersion Thermometer, D27 Distillation Receiver |
| Quantity | 1 Ea. |
| Standard Taper Joint | 19/22 |
| Joint Type | Ground Glass |
| Size | Microscale |