missing translation for 'onlineSavingsMsg'
Learn More

Shikimic Acid 97.0+%, TCI America™

Catalog No. S0038100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0038100MG 100 mg
S00381G 1 g
S00385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S0038100MG Supplier TCI America Supplier No. S0038100MG

Chemical Identifiers

CAS 138-59-0
Molecular Formula C7H9O5-
Molecular Weight (g/mol) 173.144
MDL Number MFCD00066278
InChI Key JXOHGGNKMLTUBP-HSUXUTPPSA-M
Synonym shikimic acid, shikimate, --shikimic acid, l-shikimic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-enecarboxylic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid, --shikimate, ccris 7681, unii-29ms2wi2nu, 3alpha,4alpha,5beta-trihydroxy-1-cyclohexene-1-carboxylic acid
PubChem CID 7057976
ChEBI CHEBI:36208
IUPAC Name (3R,4S,5R)-3,4,5-trihydroxycyclohexene-1-carboxylate
SMILES C1C(C(C(C=C1C(=O)[O-])O)O)O

Specifications

Melting Point 185°C
Color White
Quantity 100 mg
Formula Weight 174.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Shikimic Acid
EINECSNumber (3)-4476&(3)-4491
RTECSNumber GW4600000
TSCA No