missing translation for 'onlineSavingsMsg'
Learn More

(-)-Shikimic acid, 98%

CAS: 138-59-0 | C7H9O5- | 173.144 g/mol

Supplier:  Thermo Scientific Chemicals L0484803

 View more versions of this product

Catalog No. AAL0484803


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

(-)-Shikimic acid
184°C to 188°C
C7H9O5-
1 g
Hygroscopic
shikimic acid, shikimate, --shikimic acid, l-shikimic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-enecarboxylic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid, --shikimate, ccris 7681, unii-29ms2wi2nu, 3alpha,4alpha,5beta-trihydroxy-1-cyclohexene-1-carboxylic acid
−175° (c=2 in Water)
(3R,4S,5R)-3,4,5-trihydroxycyclohexene-1-carboxylate
7057976
174.16
138-59-0
Odorless
MFCD00066278
4782717
14,8480
JXOHGGNKMLTUBP-HSUXUTPPSA-M
C1C(C(C(C=C1C(=O)[O-])O)O)O
173.144
CHEBI:36208
98%

Chemical Identifiers

138-59-0
173.144
JXOHGGNKMLTUBP-HSUXUTPPSA-M
7057976
(3R,4S,5R)-3,4,5-trihydroxycyclohexene-1-carboxylate
C7H9O5-
MFCD00066278
shikimic acid, shikimate, --shikimic acid, l-shikimic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-enecarboxylic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid, --shikimate, ccris 7681, unii-29ms2wi2nu, 3alpha,4alpha,5beta-trihydroxy-1-cyclohexene-1-carboxylic acid
CHEBI:36208
C1C(C(C(C=C1C(=O)[O-])O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-334-2

RTECSNumber : GW4600000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only