Learn More
Justrite™ Shelves for Justrite™ Acids and Caustics Cabinets
Provide additional protection for cabinets
$598.00
Specifications
Includes Slightly raised curved edge to contain spills
View More Specs
Products 1
Catalog Number Mfr. No. Dimensions (D x W x H) Material Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (D x W x H) Material Product Type Price Quantity & Availability  
1798585A
View Documents Promotion Details
Justrite™
24195
40 x 15 in. Wood Cabinet Shelf
Each for $598.00
Only null left
Description
Description
  • Optional shelf and polyethylene tray with slighty raised curved edge to contain spills
  • Can be used in bottom sills to catch incidental spills and leaks
  • Fit select Justrite cabinets
Specifications
Specifications
Slightly raised curved edge to contain spills