Learn More
Thermo Scientific™ Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with these shelf kits for Thermo Scientific™ Ultra-Low Temperature Freezers.
Supplier: Thermo Scientific™ RSK600SD4
Description
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.
Features:
- Stainless steel racks
- Boxes
- Cell dividers
Warranty varies by country. Please contact us for details.
Specifications
| 100 | |
| Boxes (150/6 Racks) | |
| Thermo Scientific TSX and TDE Series (28.8 cu.ft.) | |
| Shelf Kit |
| Shelf Kit | |
| Stainless Steel | |
| 6 | |
| 4 Tiers |