Learn More
Thermo Scientific™ Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with these shelf kits for Thermo Scientific™ Ultra-Low Temperature Freezers.
Supplier:  Thermo Scientific™ RSK23SD4
 View more versions of this product
Catalog No. RSK23SD4
Only null left
Description
Description
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.
Features
  • Stainless steel racks
  • Boxes
  • Cell dividers
Warranty varies by country. Please contact us for details.
Specifications
Chemical Identifiers
100
Specifications
100
Boxes (100/5 Racks)
Forma 900, Forma 7000, TSE and TSD Series (23 cu.ft.)
5 sliding drawer racks and (100) 2 in. boxes with 100 cell dividers
5 Tiers
Shelf Kit
Stainless Steel
5
Shelf Kit
Product Suggestions
Product Suggestions