Learn More
Thermo Scientific™ Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with these shelf kits for Thermo Scientific™ Ultra-Low Temperature Freezers.
Supplier: Thermo Scientific™ RSK28CFSD4
Description
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.
- 6 stainless steel racks
- 150 boxes
- 100 cell dividers
Warranty
Warranty varies by country, please contact us for details
Chemical Identifiers
| 100 |
Specifications
| 100 | |
| White | |
| Stainless Steel, Plastic | |
| 6 | |
| Freezer Rack |
| Autoclavable (Racks Only) | |
| Boxes (150/6 Racks) | |
| Forma 900, Forma 7000, TSE and TSD Series (28 cu.ft.) | |
| 6 sliding drawer racks and (150) 2 in. boxes with 100 cell dividers |