missing translation for 'onlineSavingsMsg'
Learn More

IgG Sheep anti-Rabbit, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP510PMI
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP510PMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP510PMI Supplier MilliporeSigma Supplier No. AP510P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sheep Polyclonal Antibody

Sheep Anti-Rabbit IgG Antibody, HRP conjugate validated for use in western blotting.

Specifications

Antigen Sheep anti-Rabbit IgG
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate HRP
Formulation Purified sheep polyclonal in buffer containing PBS, StabilZyme™ HRP Conjugate Stabilizer, and 0.05% ProClin™ 150.
Host Species Sheep
Immunogen Rabbit IgG
Purification Method Octanoic Acid and Ammonium Sulfate cut. Depleted against Goat IgG and Rabbit IgM.
Quantity 1 mg
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8 °C protected from light in undiluted aliquots for up to 6 months
Form Purified
Show More Show Less

For Research Use Only