missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Sheep anti-Mouse, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Sheep Anti-Mouse IgG Antibody, Species absorbed: Rat IgG is an antibody against Sheep Mouse IgG for use in ELISA.
Spécifications
Spécifications
| Antigène | Sheep anti-Mouse IgG Species absorbed: Rat IgG |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Formule | Purified sheep Polyclonal serum in buffer containing PBS with 0.05% sodium azide. |
| Espèces hôtes | Sheep |
| Immunogène | Purified antibody corresponding to mouse IgG. |
| Méthode de purification | Fractionated by Octanoic Acid and Ammonium Sulfate cut and then depleted with Rat IgG column and Mouse IgA column. |
| Quantité | 1 mg |
| Primaire ou secondaire | Secondary |
| Afficher plus de résultats |
For Research Use Only