missing translation for 'onlineSavingsMsg'
Learn More

IgG Sheep anti-Human, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AC112MI
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC112MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC112MI Supplier MilliporeSigma Supplier No. AC112

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sheep Polyclonal Antibody

Sheep anti-Human IgG Antibody detects level of Sheep Human IgG and has been published and validated for use in Sheep Human IgG.

Specifications

Antigen Sheep anti-Human IgG
Applications ELISA
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified sheep polyclonal in buffer containing PBS with 0.05% sodium azide.
Host Species Sheep
Immunogen Purified human IgG
Purification Method Octanoic Acid and Ammonium Sulfate cut then depleted
Quantity 1 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C for up to 12 months.
Form Purified
Show More Show Less

For Research Use Only