missing translation for 'onlineSavingsMsg'
Learn More

Sheather's Sugar Solution, Sp. Gr. 1.250, for Flotation of Parasites, Ricca Chemical

Catalog No. 671332
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
671332 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 671332 Supplier Ricca Chemical Company Supplier No. 671332

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Sheather's Sugar Solution
Color Colorless
CAS 57-50-1, 7732-18-5, 50-00-0
CAS Min % 55.0, 42.66, 0.78
CAS Max % 57.0, 44.4, 0.81
Grade Laboratory
Concentration or Composition (by Analyte or Components) Sp. Gr. 1.250 for Flotation of Parasites
Packaging Natural Poly Bottle
pH Neutral
Quantity 1 L
Physical Form Liquid
Show More Show Less